Learn More
Safety First Aid Eyewash 3D Projecting Sign
Product Code. 31759395
Change view
Click to view available options
Color:
Green
Unit Size:
Each
Product Code. Color Unit Size
31759395 Green Each
1 options
Product Code. 31759395 Supplier Safety First Aid Supplier No. SS8004R
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Make your emergency eyewash station unmistakable, even from a distance. This Eyewash 3D Projecting Sign, 43 x 20 cm is designed for maximum visibility in busy or complex environments.
  • Mounted perpendicular to the wall, the double-sided 3D format ensures the eyewash location is visible from both directions, even down long corridors or across production floors.
  • The sign displays the standard ISO 7010 E011 eyewash icon, a white pictogram on vivid green backing, to meet UK Health Safety (Safety Signs Signals) Regulations 1996 and align with EN ISO 7010:2020 guidelines.
  • Material Installation:
    • Built from 0.7 mm rigid PVC, providing a balance of strength and lightweight design.
    • Pre-drilled for straightforward screw mounting, ideal for walls, doorframes, or posts.
    • UV-resistant ink for long-lasting visual clarity and fade protection.
  • Key Benefits:
    • Visible from multiple angles to guide people from different directions toward eyewash facilities.
    • Fast and confident response in emergencies, enhancing workplace safety.
    • Compliance-ready: fits ISO and UK signage standards, offering peace of mind to safety teams.
    • Low maintenance: simple wipe-clean surface and weather-resistant materials ensure longevity.
  • Best For:
    • Manufacturing plants and laboratories
    • Chemical handling zones and cleanrooms
    • Schools, care homes, and healthcare facilities
    • Workshops, offices, public buildings and transport hubs
Specifications
Material Plastic
Mounting Type Self-adhesive
Shape Square
Languages English
Product Type First Aid Sign
Color Green