missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Amine-PEG2-Biotin

Product Code. 11871215 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11871215 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11871215 Supplier Thermo Scientific™ Supplier No. 21346
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Biotinylate proteins or surfaces using EDC or other amine-reactive crosslinkers with this short (20.4A) biotinylation reagent containing a 2-unit polyethylene glycol spacer.

Thermo Scientific™ EZ-Link Amine-PEG2-Biotin is a biotinylation reagent that contains a 2-unit polyethylene glycol (PEG) spacer arm and a terminal primary amine for conjugation and protein labeling with EDC or other crosslinkers.

Amine-PEG2-Biotin and is a water-soluble biotin compound containing a polyethylene glycol (PEG) spacer arm and a terminal primary amine. The hydrophilic PEG spacer arm imparts water solubility that is transferred to molecules that are conjugated to the biotin compound. The PEG spacer arm also gives this reagent a long and flexible connection to minimize steric hindrance involved with binding to avidin molecules. The amine group can be reacted with the crosslinker EDC to form conjugates with carboxylates and 5' phosphates.

Highlights:

  • Use the EDC crosslinker to couple this analog to carboxyl (-COOH) groups or 5' phosphate groups
  • Water-soluble, polyethylene glycol (PEG) spacer arm
  • PEG spacer arm transfers hydrophilicity to final conjugate
TRUSTED_SUSTAINABILITY

Specifications

Product Type Amine-PEG2-Biotin
Quantity 50 mg
Content And Storage Store at 4°C.
Product Line EZ-Link
Cell Permeability Cell-Impermeant
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Reactive Moiety Amine
Solubility Water
Label or Dye Biotin
Spacer Mid-length, Pegylated
Label Type Biotin & Analogs
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.