missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ HPDP-Biotin, No-Weigh™ Format

Product Code. 15900463 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
10 x 1 mg
Unit Size:
50mg
Pack of 10
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 15900463

missing translation for 'mfr': Thermo Scientific™ A35390

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Thermo Scientific™ EZ-Link HPDP-Biotin is pyridyldithiol-activated, sulfhydryl-reactive biotinylation reagent that conjugates via a cleavable (reversible) disulfide bond to enable use in a variety of purification methods.

HPDP-Biotin is a pyridyldithiol-biotin compound for labeling protein cysteines and other molecules that contain sulfhydryl groups. This reagent specifically reacts with reduced thiols (-SH) in near-neutral buffers to form reversible disulfide bonds. HPDP-Biotin is useful for labeling and affinity-purification applications that require recovery of the original, unmodified molecule. For example, a protein can be biotinylated, allowed to bind its interactor, then captured to a streptavidin column and finally eluted and recovered by reduction of the disulfide bond with dithiothreitol.

  • Protein labeling: Biotinylate antibodies or other proteins for use in protein methods
  • Thiol-reactive: Reacts with sulfhydryls (-SH), such as the side-chain of cysteine (C)
  • Pyridyldithiol-activated: Perform reactions at pH 6.5 to 7.5 in buffers such as PBS
  • Reversible: Forms disulfide bonds, which can be cleaved using DTT or other reducing agent
  • Solubility: Must be dissolved in DMSO or DMF before further dilution in aqueous buffers
  • Medium length: Spacer arm (total length added to target) is 29.2A0
  • Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging helps prevent the loss of reagent reactivity and contamination over time by minimizing the repetitive opening and closing of the vial. The format enables use of a fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.

TRUSTED_SUSTAINABILITY

Spezifikation

Quantity 10 x 1 mg
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Room Temperature
Product Line EZ-Link, No-Weigh
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Format Solid
Label Type Biotin & Analogs

For Research Use Only. Not for use in diagnostic procedures.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt