missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Maleimide-PEG4-DBCO

Catalog No. 17611586 Alle ansehen Thermo Scientific Produkte
Ansicht ändern
Click to view available options
Quantity:
25 mg
10 x 1 mg
Packungsgröße:
25mg
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packungsgröße
17611586 25 mg 25mg
17631586 10 x 1 mg Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Catalog No. 17611586 Lieferant Thermo Scientific™ Lieferanten-Nr. C20041
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Edge
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

A sulfhydryl reactive heterobifunctional crosslinker which contains a dibenzylcyclooctyne (DBCO) group for copper-free click chemistry applications. The thiol/sulfhdryl reactive maleimide can be used to label a variety of thiol containing moieties such as free cystines in proteins, peptides, or other (bio)molecules.

  • A sulfhydryl reactive heterobifunctional crosslinker which contains a dibenzylcyclooctyne (DBCO) group for copper-free click chemistry applications
  • The thiol/sulfhdryl reactive maleimide can be used to label a variety of thiol containing moieties such as free cystines in proteins, peptides, or other (bio)molecules
  • Once a protein or (bio)molecule is DBCO labeled, it can undergo a copper-free azide-alkyne cycloaddition reaction to form a stable triazole linkage with an azide labeled coupling partner
  • Can be used to create diverse conjugates with the biorthogonal coupling between an alkyne and azide (supplied separately)
  • The coupling or crosslinking procedure is typically done in two steps:
    • First, a protein or thiol containing molecule is labeled with a DBCO group and excess crosslinker is removed
    • Second, an azide containing moiety (prepared independently) is introduced into the same system as the DBCO moiety
    • Labeling occurs under biological conditions without the introduction of any additional reagents such as a copper catalyst
  • Reagent readily dissolves in water-miscible solvents for dilution into aqueous reaction mixtures
TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Reactivity Azide-Sulfhydryl
Cleavable No
Description Maleimide-PEG4-DBCO
Molecular Weight (g/mol) 647.74
PEGylated Yes
Spacer Arm Length 29.7 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability Not determined
Shipping Condition Wet Ice
Product Line EZ-Link No-Weigh
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, DBCO
Spacer Medium (10 to 30 Å)
Form Solid
Quantity 25 mg
Solubility DMF, DMSO
Format Standard, Single-use
Water Soluble No
Product Type Crosslinker
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.