missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ NHS-PEG4 Biotinylation Kit

Product Code. 10701185 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
8 Reaction Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10701185 8 Reaction Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10701185 Supplier Thermo Scientific™ Supplier No. 21455
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Biotinylate protein amines and maximize solubility of proteins with this long (29.0A) NHS-ester biotinylation reagent containing a pegylated, water-soluble spacer arm.

Thermo Scientific™ EZ-Link NHS-PEG4-Biotin is a pegylated, water-soluble reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.

Highlights:

  • Water-soluble polyethylene glycol (PEG) spacer arm increases solubility of labeled protein
  • Prevents aggregation of antibodies better than Sulfo-NHS-LC-Biotin
  • Labels at N-terminus or lysine residues in less than 1 hour
  • Thermo Scientific No-Weigh Packaging (8 x 2mg) saves time - preserves reagent activity and prevents cross-contamination between use
TRUSTED_SUSTAINABILITY

Specifications

Description EZ-Link NHS-PEG4 Biotinylation Kit
Product Type NHS-PEG4 Biotinylation Kit
Quantity 8 Reaction Kit
Content And Storage Sufficient For: 8 labeling reactions, each with 1 to 10 mg of antibody
• EZ-Link NHS-PEG4-Biotin, 8 x 2 mg
• PBS Pack (makes 500mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 5 mL, 10 columns
• HABA Solution, 1 mL
• Avidin, 10mg

Store avidin and microtubes of biotin reagent at -20°C. Store PBS at room temperature. Store remaining kit components at 4°C.
Labeling Scale 1 to 10 mg
Product Line EZ-Link
Solubility Water
Label or Dye Biotin
Spacer Pegylated
Labeling Target Antibodies, Proteins
compliance-icons
Product Identifier
  • EZ-Link NHS-PEG4 Biotinylation Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium chloride, Sodium phosphate dibasic

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.