missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Sulfo-NHS-Biotin

Product Code. 17872556 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
10 x 1 mg
Unit Size:
50mg
Pack of 10
This item is not returnable. View return policy

Product Code. 17872556

Brand: Thermo Scientific™ A39256

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific™ EZ-Link™ Sulfo-NHS-Biotin is a short-chain, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.

Label antibodies, proteins and other molecules that have primary amines with Thermo Scientific™ EZ-Link Sulfo-NHS-Biotin, a short-chain, water-soluble biotinylation reagent.

  • Protein labeling - biotinylate antibodies to facilitate immobilization, purification or detection using streptavidin resins or probes
  • Cell surface labeling - biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
  • Amine-reactive - reacts with primary amines (-NH2), such as lysine side-chains or the amino-termini of polypeptides
  • Soluble - charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
  • Irreversible - forms permanent amide bonds; spacer arm cannot be cleaved
  • Very short - spacer arm (total length added to target) is 13.5 angstroms; it consists of the native biotin valeric acid group only.
TRUSTED_SUSTAINABILITY

Specifications

Kit Contents 10 x 1 mg
Product Type Sulfo-NHS-Biotin
Quantity 10 x 1 mg
Content And Storage Store desiccated at -20°C.
Labeling Method Chemical Labeling
Shipping Condition Qualified for Ambient or Wet Ice
Product Line EZ-Link
Cell Permeability Cell-Impermeant
Chemical Reactivity Amine
Reactive Moiety Sulfo-NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Spacer Short
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.