missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Micro Sulfo-NHS-LC-Biotinylation Kit

Product Code. 10045833 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
8 Reactions
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10045833 8 Reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10045833 Supplier Thermo Scientific™ Supplier No. 21935
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Biotinylate protein amines (e.g., lysines) with this very popular mid-length (22.4A), water-soluble, NHS-ester biotinylation reagent for antibody or cell-surface labeling.

Thermo Scientific™ EZ-Link Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.

Highlights:

  • Water-soluble analog of NHS-LC-Biotin
  • Reacts with primary amines at pH 7-9 in non-amine-containing buffers such as borate, carbonate or phosphate buffers
  • Negative charge of -SO3 group keeps biotinylation localized at the cell surface
  • Creates an amide bond
U.S. Patents 5,892,628 and 5,872,261.
TRUSTED_SUSTAINABILITY

Specifica

Description EZ-Link Micro Sulfo-NHS-LC-Biotinylation Kit
Product Type Micro Sulfo-NHS-LC-Biotinylation Kit
Quantity 8 Reactions
Content And Storage Sufficient For: 8 labeling reactions, each with 0.05–0.2 mg of antibody
• EZ-Link Sulfo-NHS-LC-Biotin, 8 x 1 mg
• PBS Pack (makes 500 mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 2 mL, 10 columns

Store biotin reagent at -20°C. Store remaining kit components at 4°C.
Labeling Scale 100 μg
Product Line EZ-Link
Solubility Water
Label or Dye Biotin
Labeling Target Antibodies, Proteins

For Research Use Only. Not for use in diagnostic procedures.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.