missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Biotinylation Kit

Product Code. 10117773 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 reactions
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10117773 10 reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10117773 Supplier Thermo Scientific™ Supplier No. 21435

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Biotinylate protein amines (e.g., lysines) with this very popular mid-length (22.4A), water-soluble, NHS-ester biotinylation reagent for antibody or cell-surface labeling.

Thermo Scientific™ EZ-Link Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.

Highlights:

  • Water-soluble analog of NHS-LC-Biotin
  • Reacts with primary amines at pH 7-9 in non-amine-containing buffers such as borate, carbonate or phosphate buffers
  • Negative charge of -SO3 group keeps biotinylation localized at the cell surface
  • Creates an amide bond
U.S. Patents 5,892,628 and 5,872,261.
TRUSTED_SUSTAINABILITY

Specifications

Description EZ-Link Sulfo-NHS-LC-Biotinylation Kit
Product Type Sulfo-NHS-LC-Biotinylation Kit
Quantity 10 reactions
Content And Storage Sufficient For: 10 labeling reactions, each with 1 to 10 mg of antibody
• EZ-Link Sulfo-NHS-LC-Biotin, 25 mg
• PBS Pack (makes 500 mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 5 mL, 10 columns
• HABA Solution, 1 mL
• Avidin, 10 mg

Store biotin and avidin reagents at -20°C. Store remaining kit components at 4°C.
Labeling Scale 1 to 10 mg
Product Line EZ-Link
Solubility Water
Label or Dye Biotin
Labeling Target Antibodies, Proteins
compliance-icons
Product Identifier
  • Sulfo-NHS-LC-Biotinylation Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium chloride, Sodium phosphate dibasic

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.