missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F(ab')2-Goat anti-Mouse IgG (H+L) Secondary Antibody, Qdot™ 605
F(ab')2-Goat Polyclonal Secondary Antibody
330.00 € - 526.00 €
Specifications
| Antigen | Mouse IgG (H+L) |
|---|---|
| Content And Storage | 4°C, store in dark |
| Applications | Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Qdot 605 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
10009372
|
Invitrogen™
Q11001MP |
200 μL |
526.00 €
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
10180723
|
Invitrogen™
Q11002MP |
100 μL |
330.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Qdot nanocrystals are composed of semi-conductor material to generate a fluorescent particle which is exceptionally bright and does not photobleach. Qdot nanocrystals paired with the correct optical filters are as much as 50 times brighter than traditional organic dyes.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
| Mouse IgG (H+L) | |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Qdot 605 | |
| Goat | |
| RUO | |
| 0.05M borate with 1M betaine and 0.05% sodium azide; pH 8.3 | |
| Secondary | |
| Purified |
| 4°C, store in dark | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| Gamma Immunoglobins Heavy and Light chains. | |
| Secondary Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title