Learn More
F(ab')2-Rabbit anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, AP, Invitrogen™
F(ab')2-Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen 31302
Code : A0
Additional Details : Weight : 0.00050kg
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31302 has been successfully used in Western blot applications. Product # 31302 reacts with heavy chains on goat IgG and with the light chains common to most goat immunoglobulins, but does not react against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human serum protein. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 0.5 mL of distilled water (0.6 mg/mL after restoration). Country of Origin: USA
Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Goat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Rabbit | |
500 μL | |
Secondary | |
4° C | |
IgG |
Western Blot | |
0.6 mg/mL | |
TBS with 15mg/mL BSA and 0.05% sodium azide; pH 8 | |
Antigen affinity chromatography | |
RUO | |
Goat | |
Lyophilized |