missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Fabric Coveralls With Hood
BIOPHARMA

Product Code. 12787186
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Pack of 20
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12787186 Large Pack of 20
12707196 Small Pack of 20
12797186 Medium Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12787186 Supplier Nitritex™ Supplier No. BDCHTL
Control Your Critical Environments
Use promo code 28022
View all promotions
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

The BioClean-D™ coverall with hood is a disposable garment featuring a front zip with protective flap, elasticated hood, back, cuffs and ankles and thumb loops to ensure a secure hold. The BioClean-D coverall provides exceptional comfort and protection, even when handling cytotoxic drugs

  • Low-linting & durable CleanTough™material
  • Thumb loops to ensure a secure hold
  • Zip with sealable cover
  • Elasticated hood, back, cuffs and ankles
  • Tested against permeation standard ASTM D6978-05
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Large
Material Polypropylene
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Zipper
Sterility Non-sterile
Resistance Isopropyl Alcohol
Back Style Elastic
Certifications/Compliance ISO 9001:2008
Description Zip front with flap cover, elasticated hood, back, cuffs and ankles
Size (Metric) Chest 990.6 to 1066.8 mm
For Use With (Application) Controlled Environments
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.