Learn More
3M™ Faceshield, Polycarbonate, Clear, 5F-11, 10 ea/Case
Product Code. 15170229 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Unit Size:
Pack of 10
Product Code. Lens Coating Unit Size
15170229 Anti-fog Pack of 10
1 options
Product Code. 15170229 Supplier 3M™ Supplier No. 7100029680
  • ONLINE EXCLUSIVE PRICE
    211.00 €
    139.62 € / Pack of 10
    Save 71.38 €
    33% Off
Estimated Shipment: 02-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
3M™ Faceshield, 5F-11 is our optical class 1, clear curved polycarbonate faceshield with anti-fog and anti-scratch coating. It’s compatible with 3M™ V5 Faceshield System for mounting on 3M™ Hard Hat Series G3000 or G22.
  • 3M™ Faceshield certified to EN 166:2001
  • Provides protection against secondary welding light
  • Anti-fog and anti-scratch coated, optical class 1, clear curved polycarbonate faceshield
  • Designed to fit 3M™ Hard Hats Series G3000 or G22 with 3M™ V5 Faceshield Mounting system
  • Snap in feature makes it quick and easy to connect to the Faceshield Holder
  • Medium energy impact, Grade B
  • Optical class 1 visor, low energy impact, B
TRUSTED_SUSTAINABILITY
Specifications
Thickness (Metric) 1.5 mm
Lens Coating Anti-fog
Product Type Faceshield
Material Polycarbonate