Learn More
Abcam Factor Xa Activity Assay Kit (Fluorometric)
Product Code. 32113153 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 x 100 Tests
Unit Size:
100 tests
Product Code. Quantity Unit Size
32113153 1 x 100 Tests 100 tests
1 options
Product Code. 32113153 Supplier Abcam Supplier No. ab3253581x100Tests
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Measure Factor X activity in 40m in with Factor Xa Activity Assay Kit AB325358. Sensitivity >1 ng. Use with Microplate (12 x 8 well strips).
Measure Factor X activity in 40m in with Factor Xa Activity Assay Kit AB325358. Sensitivity >1 ng. Use with Microplate (12 x 8 well strips).
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1 x 100 Tests