Learn More
3M™ Fahrenheit™ Unventilated Safety Goggles
Product Code. 15740028 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Unit Size:
Pack of 10
Product Code. Lens Coating Unit Size
15740028 Anti-fog Pack of 10
1 options
Product Code. 15740028 Supplier 3M™ Supplier No. 7100010410
  • ONLINE EXCLUSIVE PRICE
    230.00 €
    141.36 € / Pack of 10
    Save 88.64 €
    38% Off
View all promotions
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Fahrenheit™ Unventilated Safety Goggles have a soft crystal frame with a wide support area all around the face to enhance comfort.
  • Soft, crystal-clear PVC frame goggles with a wide support area all around the face to enhance comfort.
  • Specific channel on the side of the frame to accommodate the temple of prescription frames.
  • Model fits well with prescription glasses.
  • Special neoprene headband for good chemical resistance and easy cleaning.
  • Sealed version to protect against liquid droplets and large dust particles.
  • Anti-Fog coating to help reduce lens fogging.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ETIM Class 5.0, EC002098, CE Approved , EN 166: 2001
Lens Coating Anti-fog
Description Sealed
Product Type Safety Goggles
For Use With (Application) Construction, food industry
Frame Color Clear
Headband Material Neoprene
Ventilation Details Unventilated
Material PVC
Lens Material Acetate
Frame Type Soft Frame