missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ Sterile Sample Containers

Product Code. 10100881 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
110 mL
220 mL
Closure Included:
No
Yes
Unit Size:
Pack of 100
Pack of 500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Included unitSize
10100881 220 mL Yes Pack of 100
10153862 220 mL No Pack of 500
10002922 110 mL No Pack of 500
10190871 110 mL Yes Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10100881 Supplier Falcon™ Supplier No. 354015
  • ONLINE EXCLUSIVE PRICE
    117.00 €
    108.71 € / Pack of 100
    Save 8.29 €
    7% Off
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Made of clear polypropylene for strength and visibility. Falcon™ Sterile Sample Containers are ideal for specimen collection.

  • Sterile, disposable polypropylene containers provide secure sample containment
  • Available in 4 1/2 oz. (110 mL) and 8 oz. (220 mL) sizes both with and without matching lids
  • Feature molded-in graduations for easy measurements
  • Graduated in ounces and milliliters to provide convenient, single-use, timesaving containers for collection, transportation, and storage of a wide variety of specimens
  • Inert and chemically resistant to commonly used laboratory reagents at room temperature
  • Features

    Sterile
  • 4 1/4 oz. graduations from 1/2 oz. to 4 1/2 oz. in 1/4 oz. increments and from 20 mL to 110 mL in 10 mL increments
  • 8 oz. size graduated from 1/4 oz. to 8 oz. in 1/4 oz. increments and from 20 mL to 220 mL in 10 mL increments
TRUSTED_SUSTAINABILITY

Specifications

Closure Included Yes
Graduated Yes
Material Polypropylene
Sterility Sterile
Capacity (Metric) 220 mL
No. per Case 100/Cs.
No. per Pack 1/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.