missing translation for 'onlineSavingsMsg'
Learn More

Falipamil, TRC

Product Code. 30138593
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138593 50 mg 50mg
30135700 100 mg 100mg
30006763 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138593 Supplier Toronto Research Chemicals Supplier No. TRCF10111650MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77862-92-1
Molecular Formula C24H32N2O5
Molecular Weight (g/mol) 428.52
Synonym 2-[3-[[2-(3,4-Dimethoxyphenyl)ethyl]methylamino]propyl]-2,3-dihydro-5,6-dimethoxy-1H-isoindol-1-one, AQ-A 39;
SMILES COc1ccc(CCN(C)CCCN2Cc3cc(OC)c(OC)cc3C2=O)cc1OC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H32N2O5/c1-25(12-9-17-7-8-20(28-2)21(13-17)29-3)10-6-11-26-16-18-14-22(30-4)23(31-5)15-19(18)24(26)27/h7-8,13-15H,6,9-12,16H2,1-5H3
Formula Weight 428.2311
Chemical Name or Material Falipamil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.