Product Code. 15662670
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
15662670
4 x 2.5 L
Glass Bottle
Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
15662670
Supplier
Fluka
Supplier No.
245384X2.5L
610.00 € 572.59 € / Pack of 4 Save 37.41 € 6% Off
Estimated Shipment: 19-03-2027Log in to see stock.
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
DIN 51635, bp 65-95 °C
Solstice has over 200 years of history in producing high-purity chemicals It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Melting Point
-30°C
CAS
101316-46-5
Density
0.690 to 0.705g/mL at 15°C
Quantity
4 x 2.5 L
Flash Point
−26°C (−14.8°F)
Packaging
Glass Bottle
MDL Number
MFCD00081849
UN Number
UN1268
Boiling Point
65°C to 95°C
Chemical Name or Material
Fam Benzine
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?