missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM™ Dye-Labeled Anti-miR™ Negative Control #1

Product Code. 10457684
Change view
Click to view available options
Quantity:
5 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10457684 5 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10457684 Supplier Invitrogen™ Supplier No. AM17012
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones

FAM™ Dye-Labeled Anti-miR™ Negative Control #1

A FAM™ dye–labeled synthetic miRNA inhibitor for miRNA functional analysis. 5 nmol is supplied, purified and ready to transfect. Invitrogen™ Anti-miR™ miRNA Inhibitors are chemically modified, single-stranded nucleic acids designed to specifically bind to and inhibit endogenous microRNA (miRNA) molecules. These ready-to-use inhibitors can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. FAM™ Dye Labeled Anti-miR™ Negative Control #1 is a random sequence Anti-miR molecule that has been extensively tested in human cell lines and tissues and validated to produce no identifiable effects on known miRNA function. It is used for monitoring transfection efficiency in transfection experiments using Anti-miR miRNA Inhibitors. The fluorescent label enables direct observation of cellular uptake, distribution, and localization of the control. The molecule is labeled at its 5' end and has the same oligonucleotide sequence as unlabeled Anti-miR™ miRNA Inhibitor Negative Control #1 (SKU #AM17010).

Accessory Products:
Also available are Anti-miR™ miRNA Inhibitors (SKU #AM17000 and #AM17001). To select your miRNA sequence and order, please visit the GeneAssist™ miRNA Workflow Builder. Cy™3 dye-labeled Anti-miR™ Negative Controls (SKU #AM17011) are available. The Pre-miR™ miRNA Starter Kit (SKU #AM1540) can be used to optimize transfection of Pre-miR™ miRNA Precursors.
TRUSTED_SUSTAINABILITY

Especificaciones

Content And Storage Anti-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C.
Control Type Negative Control
RNAi Type miRNA Inhibitor
Species Human, Mouse, Rat
For Use With (Application) miRNA & Non-Coding RNA Analysis
Label or Dye FAM
Product Line Ambion, Anti-miR, FAM
Product Type miRNA Inhibitor
Purity HPLC
Quantity 5 nmol
Shipping Condition Dry Ice
Mostrar más Mostrar menos

For Research Use Only. Not for use in diagnostic procedures.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.