missing translation for 'onlineSavingsMsg'
Learn More

Famprofazone, TRC

Product Code. 30138544
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138544 100 mg 100mg
30134925 25 mg 25mg
17913015 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138544 Supplier Toronto Research Chemicals Supplier No. TRCF102350100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22881-35-2
Molecular Formula C24 H31 N3 O
Molecular Weight (g/mol) 377.52
Synonym 3H-Pyrazol-3-one, 1,2-dihydro-1-methyl-4-(1-methylethyl)-5-[[methyl(1-methyl-2-phenylethyl)amino]methyl]-2-phenyl-, 3-Pyrazolin-5-one, 4-isopropyl-2-methyl-3-[[methyl(α-methylphenethyl)amino]methyl]-1-phenyl- (8CI), Famprofazone
IUPAC Name 1-methyl-5-[[methyl(1-phenylpropan-2-yl)amino]methyl]-2-phenyl-4-propan-2-ylpyrazol-3-one
SMILES CC(C)C1=C(CN(C)C(C)Cc2ccccc2)N(C)N(C1=O)c3ccccc3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C24H31N3O/c1-18(2)23-22(17-25(4)19(3)16-20-12-8-6-9-13-20)26(5)27(24(23)28)21-14-10-7-11-15-21/h6-15,18-19H,16-17H2,1-5H3
Formula Weight 377.2467
Chemical Name or Material Famprofazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.