missing translation for 'onlineSavingsMsg'
Learn More

Farnesol, 96%, mixture of isomers

Product Code. 10366650 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10366650 100 mL Glass bottle 100mL
10356650 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10366650 Supplier Thermo Scientific Supplier No. 119121000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4602-84-0 | C15H26O | 222.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4602-84-0
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.37
MDL Number MFCD00002918,MFCD00002918,MFCD00002918,MFCD00002918
InChI Key CRDAMVZIKSXKFV-YFVJMOTDSA-N
Synonym e,z-farnesol, 2-trans,6-cis-farnesol, 2e,6z-farnesol, trans,cis-farnesol, farnesol, 2e,6z-3,7,11-trimethyldodeca-2,6,10-trien-1-ol, farnesyl alcohol, e,z-3,7,11-trimethyl-2,6,10-dodecatrien-1-ol, 2-trans,6-cis-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
PubChem CID 1549109
ChEBI CHEBI:35966
IUPAC Name (2E,6Z)-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CO

Specifications

Color Colorless to Yellow
Density 0.8865g/mL
Boiling Point 149°C (4.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (sum of geometric isomers) (GC)
Packaging Glass bottle
Refractive Index 1.4870 to 1.492
Quantity 100 mL
Beilstein 01, III, 2040
Merck Index 15, 3975
Specific Gravity 0.8865
Solubility Information Solubility in water: immiscible
Formula Weight 222.37
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Farnesol, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.