missing translation for 'onlineSavingsMsg'
Learn More

Farnesyl acetate, mixture of isomers, 96%

Product Code. 11428647
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428647 100 g 100g
11418647 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428647 Supplier Thermo Scientific Alfa Aesar Supplier No. A19778.22
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 29548-30-9 | C17H28O2 | 264.409 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29548-30-9
Molecular Formula C17H28O2
Molecular Weight (g/mol) 264.409
MDL Number MFCD00036516
InChI Key ZGIGZINMAOQWLX-NCZFFCEISA-N
Synonym farnesyl acetate, farnesol acetate, all-trans-farnesyl acetate, unii-d5zj1foc2i, trans,trans-farnesyl acetate, d5zj1foc2i, 3,7,11-trimethyl-2,6,10-dodecatrienyl acetate, 2e,6e-farnesyl acetate, 2,6,10-dodecatrien-1-ol, 3,7,11-trimethyl-, acetate, e,e, 2e,6e-3,7,11-trimethyldodeca-2,6,10-trien-1-yl acetate
PubChem CID 638500
IUPAC Name [(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienyl] acetate
SMILES CC(=CCCC(=CCCC(=CCOC(=O)C)C)C)C

Specifications

Density 0.91
Flash Point 95°C (203°F)
Refractive Index 1.477
Quantity 100 g
Beilstein 1784823
Formula Weight 264.41
Percent Purity 96%
Chemical Name or Material Farnesyl acetate, mixture of isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.