missing translation for 'onlineSavingsMsg'
Learn More

Farnesylacetone, mixture of isomers, 97%

Product Code. 11479506
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479506 250 g 250g
11469506 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479506 Supplier Thermo Scientific Alfa Aesar Supplier No. A19234.30
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 762-29-8 | C18H30O | 262.437 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762-29-8
Molecular Formula C18H30O
Molecular Weight (g/mol) 262.437
MDL Number MFCD00036517
InChI Key LTUMRKDLVGQMJU-IUBLYSDUSA-N
Synonym farnesyl acetone, 5e,9e-6,10,14-trimethylpentadeca-5,9,13-trien-2-one, farnesylacetone, e,e-farnesylacetone, unii-3s0g4n267h, e,e-6,10,14-trimethylpentadeca-5,9,13-trien-2-one, trans,trans-farnesylacetone, 5,9,13-pentadecatrien-2-one, 6,10,14-trimethyl-, e,e, 2,6,10-trimethyl-2,6,10-pentadecatrien-14-one, 5,9,13-pentadecatriene-2-one, 6,10,14-trimethyl
PubChem CID 1711945
ChEBI CHEBI:67252
IUPAC Name (5E,9E)-6,10,14-trimethylpentadeca-5,9,13-trien-2-one
SMILES CC(=CCCC(=CCCC(=CCCC(=O)C)C)C)C

Specifications

Density 0.88
Flash Point >110°C (230°F)
Odor Pleasant
Refractive Index 1.481
Quantity 250 g
Beilstein 1781239
Formula Weight 262.44
Percent Purity 97%
Chemical Name or Material Farnesylacetone, mixture of isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.