missing translation for 'onlineSavingsMsg'
Learn More

MP BIOMEDICALS FAST DNA Erase 500 mL

Product Code. 31874170 Shop All MP Biomedicals Products
Change view
Click to view available options
Volume (Metric):
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
31874170 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31874170 Supplier MP BIOMEDICALS Supplier No. 112463500
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Rapid DNA decontamination in just 10 minutes. Effective across common lab surfaces with a simple, efficient workflow, ideal for maintaining clean PCR workstations.

  • FAST DNA Erase is a ready-to-use, clear, colorless liquid designed to eliminate DNA contamination from common laboratory surfaces, including countertops, plastics, stainless steel, and glass.
  • Application Notes: FAST DNA Erase is intended to decontaminate laboratory surfaces, apparatus, and plastic and glass vessels.
  • Usage Statement: Unless otherwise specified, MP Biomedicals' products are intended for research or further manufacturing use only and are not for direct human use.
  • Key Application: Removal of DNA contamination

Specifications

Color-coded No
Form Liquid Spray
Scent Unscented
Sterility Non-sterile
Volume (Metric) 500 mL
compliance-icons
Product Identifier
  • FAST DNA Erase
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.