missing translation for 'onlineSavingsMsg'
Learn More

Fast Garnet GBC Salt, TRC

Product Code. 30406966
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406966 100 mg
30396022 1 g
30401154 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406966 Supplier Toronto Research Chemicals Supplier No. TRCF101890100MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-89-3
Molecular Formula C14H13N4 . HO4S
Molecular Weight (g/mol) 334.35
Synonym Garnet GBC Salt, 2-Methyl-4-(o-tolylazo)benzenediazonium Hydrogen Sulfate, 4-(o-Tolylazo)-o-toluenediazonium Hydrogen Sulfate (6CI)
IUPAC Name hydrogen sulfate;2-methyl-4-[(E)-(2-methylphenyl)diazenyl]benzenediazonium
SMILES Cc1ccccc1N=Nc2ccc([N+]#N)c(C)c2.OS(=O)(=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H13N4.H2O4S/c1-10-5-3-4-6-14(10)18-17-12-7-8-13(16-15)11(2)9-12;1-5(2,3)4/h3-9H,1-2H3;(H2,1,2,3,4)/q+1;/p-1
Formula Weight 334.0736
Chemical Name or Material Fast Garnet GBC Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.