missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fast Green FCF

Product Code. 11497837
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
missing translation for 'unitSize'
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11497837

missing translation for 'mfr': Thermo Scientific Chemicals A16520.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 2353-45-9 | C37H34N2Na2O10S3 | 808.84 g/mol

Stain for showing collagen, muscle, cytoplasm and cells in mammalian tissuesFast Green FCF is used as a quantitative stain for histones and protein stain in electrophoresis. It is also used as a food dye, which is poorly absorbed by intestine. It is used for tinned green peas and other vegetables, jellies, sauces, fish and desserts. It is considered as a replacement of Light Green SF yellowish in Masson's trichrome. Further, it is utilized in tissue stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Stain for showing collagen, muscle, cytoplasm and cells in mammalian tissuesFast Green FCF is used as a quantitative stain for histones and protein stain in electrophoresis. It is also used as a food dye, which is poorly absorbed by intestine. It is used for tinned green peas and other vegetables, jellies, sauces, fish and desserts. It is considered as a replacement of Light Green SF yellowish in Masson′s trichrome. Further, it is utilized in tissue stain.

Solubility
Soluble in water, glycerin, glycol and ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2353-45-9
Molecular Formula C37H34N2Na2O10S3
Molecular Weight (g/mol) 808.84
MDL Number MFCD00013053
InChI Key XJBPDZVCYTYRIN-UHFFFAOYSA-L
Synonym C.I. 42053
PubChem CID 16887
IUPAC Name disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate
SMILES [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O

Specifications

Melting Point ∼290°C (decomposition)
Beilstein 5718212
Merck Index 14,3941
Solubility Information Soluble in water,glycerin,glycol and ethanol.
Formula Weight 808.86
Sensitivity Hygroscopic
Quantity 5 g
Chemical Name or Material Fast Green FCF
compliance-icons
Product Identifier
  • Fast Green FCF
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : C.I. Food Green 3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.