missing translation for 'onlineSavingsMsg'
Learn More

Fast Green FCF, Fisher BioReagents

Product Code. 11443054
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443054 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11443054 Supplier Fisher BioReagents Supplier No. BP12310
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Fast Green FCF is a protein stain for collagenous tissue.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 290°C
CAS 2353-45-9
Assay Percent Range ≥90.0 %
Molecular Formula C37H34N2Na2O10S3
MDL Number MFCD00013053
Merck Index 15, 3978
Synonym FD & C Green No. 3
Identification Pass Test
InChI Key XJBPDZVCYTYRIN-UHFFFAOYSA-L
SMILES [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
IUPAC Name disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate
Molecular Weight (g/mol) 808.84
PubChem CID 16887
Lambda Max 620 to 624nm
Formula Weight 808.85
Percent Purity ≥90.0%
Packaging Glass Bottle
Color Index No. 42053
Color Red
Quantity 10 g
Physical Form Powder
Chemical Name or Material Fast Green FCF
Show More Show Less
compliance-icons
Product Identifier
  • Fast Green FCF
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.