missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Fast MicroSEQ™ 500 16S rDNA PCR Kit

Product Code. 30000675 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000675 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000675 Supplier Applied Biosystems™ Supplier No. 4370489
  • ONLINE EXCLUSIVE PRICE
    1660.00 €
    1598.00 € / Pack of 1
    Save 62.00 €
    3% Off
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Fast MicroSEQ™ 500 16S rDNA PCR kit utilizes new Fast technology and provides bacterial identifications from colony to results in as little as 5 hours using the Veriti™ 96-well Thermal Cycler.

The Fast MicroSEQ™ 500 16S rDNA PCR kit utilizes new Fast technology and provides bacterial identifications from colony to results in as little as 5 hours using the Veriti™ 96-well Thermal Cycler. This kit is also compatible with the GeneAmp™ PCR System 9700. The PCR-amplified products that are generated using this kit can be sequenced using the MicroSEQ™ 500 16S rDNA Seqencing Kit. The two kits together permit the amplification and sequencing of 500 base pairs of the 16S rRNA gene. These sequences can then be compared to the MicroSEQ™ ID Analysis Software 16S rDNA 500 Library to obtain an identity match. This kit does not include a protocol or a Quick Reference Card.

  • Sequence-based measurement ensures you get reproducible results in under 24 hours.
  • Growth requirement-independent measurement enables you to identify bacteria using objective criteria.
  • Universal domain-level primers eliminate up-front optimization.
  • MicroSEQ™ ID Analysis Software ensures accurate species identification.
  • Customizable libraries give you the flexibility to create new libraries and search validated libraries that contain the most frequently encountered organisms.
  • Requires minimal training-same work flow as the MicroSEQ™ D2 LSU fungal kit.

The MicroSEQ™ ID System: A Complete, Integrated Solution
Streamlining every step in microbial identification, Applied Biosystems™ MicroSEQ™ System combines the advantages of MicroSEQ™ ID Software with easy PrepMan Ultra sample preparation reagents and protocols, MicroSEQ™ bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To ensure optimal performance and system-wide integration, we develop and test all components of the system together.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each kit contains enough PCR Master Mix to perform 50 PCR amplifications, 5 negative control assays, and 5 positive control assays. One tube of positive control E. coli DNA, and one of water (for use as a negative control) are also included.

Store at -15°C to -25°C upon reciept. After first use, store at 2°C to 8°C in a PCR Clean Room.
Detection Method Primer-probe
GC-Rich PCR Performance Low
Reaction Speed Fast
Technique SBT (Sequence-Based Typing)
For Use With (Equipment) Veriti Thermal Cycler
Libraries Customizable, Bacterial Library (> 2000 species)
Product Line MicroSEQ
Product Type rDNA PCR Kit
Quantity 1 kit
Shipping Condition Dry Ice
Software MicroSEQ™ ID Analysis Software
Target 1st 500 bp of 16S rRNA gene
Target Organism Class Bacteria
For Use With (Application) Standard PCR
Sample Type DNA (Genomic)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.