missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Fast MicroSEQ™ D2 Fungal rDNA PCR Kit

Product Code. 30000669 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000669 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000669 Supplier Applied Biosystems™ Supplier No. 4382397
  • ONLINE EXCLUSIVE PRICE
    1355.00 €
    1250.00 € / Pack of 1
    Save 105.00 €
    7% Off
 Restricted Item
Estimated Shipment: 22-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This kit is the PCR component of the FAST MicroSEQ Fungal Identification System, with enough reagents for 50 PCR amplifications of fungal DNA unknowns.

This kit is the PCR component of the FAST MicroSEQ™ Fungal Identification System, with enough reagents for 50 PCR amplifications of fungal DNA unknowns. The sequencing component of the kit is required for species identification. Together, the two kits provide an easy-to-use DNA sequence-based method to identify most fungi.

  • Sequence-based measurement ensures you get reproducible results in less than 24 hours.
  • Growth requirement-independent measurement enables you to identify fungi using objective criteria.
  • Universal domain-level primers eliminate up-front optimization.
  • MicroSEQ™ ID Analysis Software ensures accurate species identification.
  • Customizable libraries give you the flexibility to create new libraries and search validated libraries that contain the most frequently encountered organisms.
  • Requires minimal training; same work flow as the MicroSEQ™ Full Gene 16S rDNA Bacterial Identification Kit.

Specific and Easy to Use
Based on phylogenetic analysis of the D2 region of large-subunit ribosomal RNA gene sequences, this easy-to-use kit enables you to identify even the most difficult fungi, yeast, and oomycete plant pathogens. To ensure accurate and specific results, the kit's software compares your samples to a database of D2 sequences from over 1000 validated type species.

The MicroSEQ™ ID System: A Complete, Integrated Solution
Streamlining every step in microbial identification, Applied Biosystems™ MicroSEQ™ System combines the advantages of MicroSEQ™ ID Software with easy PrepMan Ultra sample preparation reagents and protocols, MicroSEQ™ bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To ensure optimal performance and system-wide integration, we develop and test all components of the system together.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The kit contains enough reagent for 50 identifications:
• PCR Master Mix (store at 2°C to 8°C in PCR clean room)
• Positive PCR Control DNA (2°C to 8°C in PCR clean room)
• Negative PCR Control (2°C to 8°C in PCR clean room)
Detection Method Primer-probe
GC-Rich PCR Performance Low
Reaction Speed Fast
Technique SBT (Sequence-Based Typing)
For Use With (Equipment) GeneAmp 9700, Veriti Thermal Cycler, 9800 Fast Thermal Cycler
Libraries Fungal Library (> 1100 species), Customizable
Product Line MicroSEQ
Product Type rDNA PCR Kit
Quantity 1 Kit
Shipping Condition Dry Ice
Software MicroSEQ™ ID Analysis Software
Target D2 region of rRNA gene
Target Organism Class Fungi
Target Specificity Target-Specific
For Use With (Application) Standard PCR
Sample Type DNA (Genomic)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.