missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ FastDigest Buffer (10X)
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Quantity:
5 x 1 mL
Dimensione della confezione:
Pack of 10
Descrizione
The 10X FastDigest Buffer is a proprietary digestion buffer which supports 100% activity of all FastDigest restriction enzymes. DNA/RNA modifying enzymes, such as ligases, phosphatases, kinases and mesophilic DNA polymerases also have 100% activity in FastDigest Buffer. Therefore, enzymes used in downstream applications can be directly added to the FastDigest reaction mix
Benefits:
Double and multiple digestion in a single buffer with any combination of enzymes.
No sequential digestions or buffer changes needed.
Benefits:
Specifica
Specifica
| Concentration | 10X |
| Quantity | 5 x 1 mL |
| Product Type | Digestion Buffer |
| Research Category | Traditional Cloning |
| Product Line | FastDigest |
For Research Use Only. Not for use in diagnostic procedures.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto