missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ FastGro Synthetic, Animal Free, Chemically Defined FBS Replacement

Product Code. 16277721 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16277721 100 mL 100mL
16329855 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16277721 Supplier MP Biomedicals™ Supplier No. 092640049
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

FastGro allows culturing cells in vitro without the use of serum and without any animal or human derived compound.

FastGro allows culturing cells in vitro without the use of serum and without any animal or human derived compound. FastGro is a fully defined additive, avoiding the need for serum, for a wide range of cell culture practices. Since FastGro is chemically defined it avoids lot-to-lot variations. All components in FastGro are highly purified and identified chemical compounds.

Culturing cells in a FastGro enriched medium represents a number of benefits as compared to conventional FBS enriched media.

  • Due to its chemically defined nature there are no lot-to-lot variations. This skips the need for testing new batches of serum.
  • There is no interference of unknown serum compounds in your research.
  • Since it doesn't contain hormones nor cytokines it doesn't bias your research data.
  • FastGro is save in use, since it doesn't contain animal or human derived materials.
  • It can be stored in the refrigerator. No need for thawing before use.

Specifications

Content And Storage Store at 2 to 8°C
Form Liquid
Product Type FBS Replacement Serum
Quantity 100 mL
Sterility Sterile
Grade Cell Biology
Tested For Liver epithelial cells, HaCaT, Primary human hepatocytes, Hybridoma, VERO, HEK293, CHO, HEP G2, HeLa, Human keratinocytes
For Use With (Application) Cell Culture
compliance-icons
Product Identifier
  • FastGro Synthetic, animal free, chemically defined FBS
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.