missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals FastPrep-96™ PCR Plate Holders for DNA Shearing

Product Code. 31114748 Shop All MP Biomedicals Products
Change view
Click to view available options
Product Type:
Plate Holder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type
31114748 Plate Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31114748 Supplier MP Biomedicals Supplier No. 116010540
  • ONLINE EXCLUSIVE PRICE
    31.00 €
    29.40 € / 1 pairs
    Save 1.60 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Polypropylene deep well plates designed for high-throughput DNA shearing using the FastPrep-96™ instrument.

  • Set of two 96-well deep well plates made of durable polypropylene for reliable performance
  • Used as plate holders to mount a second PCR plate where DNA is loaded for shearing purposes
  • Provides an ideal and efficient setup for consistent and reproducible shearing of HMW DNA into smaller fragments
  • Supports library preparation for long-read sequencing platforms such as PacBio™
  • Enables high-throughput, efficient, uniform, and consistent DNA shearing across the entire plate
  • Allows processing of up to 192 samples in just a few minutes for improved throughput
  • Fits perfectly and securely within the FastPrep-96™ 2 x 96 deep well plate holder
  • Application Notes: For use with the FastPrep-96 homogenizer for high-throughput DNA shearing prior to library preparation using long-read sequencing platforms like PacBio™
  • Usage Statement: Research Use Only (RUO)

Specifications

Product Type Plate Holder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.