missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FastRuler Ultra Low Range DNA Ladder, Ready-to-Use, 10-200 bp Promotion

Product Code. 15787546 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 to 333 Applications
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15787546 50 to 333 Applications Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15787546 Supplier Thermo Scientific™ Supplier No. SM1233

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Perform fast sizing and approximate quantification of dsDNA in high throughput gels and conventional agarose gels with Thermo Scientific™ FastRuler™ DNA Ladders.

 

FastRuler Ultra Low Range DNA Ladder is specifically designed for fast sizing and approximate quantification of double-stranded DNA in 48 well (or 96-well) high throughput gels, as well as in conventional agarose gels. The ladder is a mixture of five blunt-ended chromatography-purified individual DNA fragments that are easily resolved in a short separation distance (10 to 20 mm) after an 8 to 14 minutes run on the appropriate agarose gel.

The ladder is especially useful for electrophoresis of PCR products.

FastRuler Ultra Low Range DNA Ladder, ready-to-use contains dephosphorylated DNA fragments and is ideal for forward 5-end labeling with T4 Polynucleotide Kinase (see the Certificate of Analysis for the detailed labeling protocol).

Highlights

  • Fast separation (8 to 14 min)
  • Short separation distance (10 to 20 mm)
  • Sharp bands, easy-to-remember fragment sizes and quantities
  • Ready-to-use -premixed with loading dye
  • Supplied with loading dye solution for sample DNA 
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Concentration 6X Loading Dye
Form Liquid
Kit Contents 6X Orange DNA Loading Dye, 1mL
Quantity 50 to 333 Applications
Product Type DNA Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.