missing translation for 'onlineSavingsMsg'
Learn More

FCS Express™ Flow Cytometry Analysis Software

Product Code. 17163249
Change view
Click to view available options
Quantity:
1 license
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17163249 1 license Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17163249 Supplier Invitrogen Supplier No. A48515
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

In collaboration with De Novo Software, Thermo Fisher Scientific offers FCS Express Flow Cytometry Analysis Software. FCS Express software gets you from raw data to easily understandable, beautifully formatted, presentation-ready results more easily and in less time than any other flow cytometry software.

In collaboration with De Novo Software, Thermo Fisher Scientific offers FCS Express Flow Cytometry Analysis Software. FCS Express software gets you from raw data to easily understandable, beautifully formatted, presentation-ready results more easily and in less time than any other flow cytometry software. This product consists of a 1-year license for FCS Express software obtained via digital download.

FCS Express software features include:

  • Fully integrated analysis, statistics, graphing and reporting
  • Microsoft Office™ inspired interface that makes learning easy, while integrated spreadsheets and direct high-resolution exports prevent you from using multiple software tools to achieve a final, publication-ready result
  • Full support for Attune instruments that helps ensure a seamless transition from instrument acquisition to final result
  • Dedicated and knowledgeable support team with <24 hr response times

TRUSTED_SUSTAINABILITY

Specifica

Content And Storage 1 FCS Express software license digital download. Store at room temperature.
Product Type Data Analysis software
Quantity 1 license
Shipping Condition Room Temperature
Software Type Flow software

For Research Use Only. Not for use in diagnostic procedures.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.