missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Febuxostat, 98%

Product Code. 15791769 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15791769 5 g 5g
15781769 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15791769 Supplier Thermo Scientific Chemicals Supplier No. 461580050
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 144060-53-7 | C16H16N2O3S | 316.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144060-53-7
Molecular Formula C16H16N2O3S
Molecular Weight (g/mol) 316.38
MDL Number MFCD00871598
InChI Key BQSJTQLCZDPROO-UHFFFAOYSA-N
IUPAC Name 2-[3-cyano-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC(C)COC1=CC=C(C=C1C#N)C1=NC(C)=C(S1)C(O)=O

Specifications

Melting Point 209.0°C
Assay Percent Range 97.5 % min. (HPLC)
Quantity 5 g
Formula Weight 316.38
Chemical Name or Material Febuxostat

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.