missing translation for 'onlineSavingsMsg'
Learn More

Felodipine, MedChemExpress

Product Code. 16464327
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464327 50 mg 50mg
16428448 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16464327 Supplier Medchem Express Supplier No. HYB0309
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Felodipine, a dihydropyridine, is a potent, vasoselective calcium channel antagonist. Felodipine lowers blood pressure (BP) by selective action on vascular smooth muscle, especially in the resistance vessels. Felodipine, an anti-hypertensive agent, induces autophagy. Felodipine can cross the blood-brain barrier.

Felodipine, a dihydropyridine, is a potent, vasoselective calcium channel antagonist. Felodipine lowers blood pressure (BP) by selective action on vascular smooth muscle, especially in the resistance vessels. Felodipine, an anti-hypertensive agent, induces autophagy. Felodipine can cross the blood-brain barrier.

Specifications

Chemical Name or Material Felodipine
CAS 72509-76-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H19Cl2NO4
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (260.25 mM; Need ultrasonic)
SMILES O=C(C1=C(C)NC(C)=C(C(OC)=O)C1C2=CC=CC(Cl)=C2Cl)OCC
Molecular Weight (g/mol) 384.25
Formula Weight 384.25
Percent Purity 98.93%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.