missing translation for 'onlineSavingsMsg'
Learn More

Felodipine

Product Code. 15443819
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443819 50 mg 50mg
15423819 100 mg 100mg
15433819 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15443819 Supplier Thermo Scientific Alfa Aesar Supplier No. J61195.MF
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A dihydropyridine calcium channel blocker | CAS: 72509-76-3 | C18H19Cl2NO4 | 384.253 g/mol

Felodipine is used as a dihydropyridine calcium channel blocker. It displays high vascular selectivity; lowers arterial blood pressure without altering cardiac contractility. Antihypertensive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Felodipine is used as a dihydropyridine calcium channel blocker. It displays high vascular selectivity; lowers arterial blood pressure without altering cardiac contractility. Antihypertensive.

Solubility
Soluble in DMSO. Insoluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72509-76-3
Molecular Formula C18H19Cl2NO4
Molecular Weight (g/mol) 384.253
MDL Number MFCD00868316
InChI Key RZTAMFZIAATZDJ-UHFFFAOYSA-N
Synonym felodipine, plendil, flodil, munobal, renedil, dl-felodipine, perfudal, splendil, feloday, prevex
PubChem CID 3333
ChEBI CHEBI:585948
IUPAC Name 5-O-ethyl 3-O-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C(C1C2=C(C(=CC=C2)Cl)Cl)C(=O)OC)C)C

Specifications

Color Yellow
Quantity 50 mg
Merck Index 14,3953
Solubility Information Soluble in DMSO; Insoluble in water
Formula Weight 384.25
Physical Form Crystalline Powder
Chemical Name or Material Felodipine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.