Learn More
Value Plastics Female Luer to 500 Series Barb
Product Code. 30053447 Shop All Cole-Parmer Products
Change view
Click to view available options
Inlet:
Female Luer x Hose Barb
Unit Size:
Pack of 25
Product Code. Inlet Unit Size
30053447 Female Luer x Hose Barb Pack of 25
1 options
Product Code. 30053447 Supplier Value Plastics Supplier No. FTLLSB0136005
  • ONLINE EXCLUSIVE PRICE
    23.95 €
    21.52 € / Pack of 25
    Save 2.43 €
    10% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Inlet Female Luer x Hose Barb
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.