Learn More
Ibidi Female Luer Lock Connector
Click to view available options
:
Female Luer Lock Connector
Description
- Suitable for male Luer or Luer Lock adapters
- natural Polypropylene
- sterilized
Specifications
Specifications
| Material | Polypropylene |
| For Use With | Male Luer Adapter, Male Luer Lock Adapter |
| Packaging Quantity | 25 Pk. |
| Sterility | Sterile |
| Product Type | Female Luer Tubing Connector |