Description
Luer fittings allow for a simple yet secure twist-lock connection. Ideal for capping off fluid transfer system. Thread design provides a secure closure—preventing contamination
Specifications
Specifications
| Certifications/Compliance | Non-DEHP;Non-Phthalate;RoHS |
| Packaging Quantity | 25/Pk. |
| Inlet | Female Luer Thread, Cap |
| Material (Seal) | White Nylon |