Learn More
Value Plastics Female Luer Thread Style Caps
Product Code. 30053116 Shop All Cole-Parmer Products
Change view
Click to view available options
Material (Seal):
Clear Polycarbonate
White Nylon
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material (Seal) unitSize
30053116 Clear Polycarbonate Pack of 25
30052968 White Nylon Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30053116 Supplier Value Plastics Supplier No. 5012041
Only null left
Add to basket
Add to basket
Seal luer ports with a secure and leak-proof connection
Luer fittings allow for a simple yet secure twist-lock connection. Ideal for capping off fluid transfer system. Thread design provides a secure closure—preventing contamination
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS;FDA
Packaging Quantity 25/Pk.
Inlet Female Luer Thread, Cap
Material (Seal) Clear Polycarbonate