Learn More
Value Plastics Female Luer Thread Union Fittings
Product Code. 30053660 Shop All Cole-Parmer Products
Change view
Click to view available options
Material (Seal):
Polycarbonate
Polypropylene
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material (Seal) unitSize
30053660 Polycarbonate Pack of 25
30053066 Polypropylene Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30053660 Supplier Value Plastics Supplier No. 5012066
Only null left
Add to basket
Add to basket
Luer fittings allow for a simple yet secure connection
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standards
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS;FDA
Packaging Quantity 25/Pk.
Inlet Female Luer Thread
Material (Seal) Polycarbonate