Learn More
Fendiline Hydrochloride, Tocris Bioscience™
Inhibits KRas localization to the plasma membrane; also L-type calcium channel blocker
Brand: Tocris Bioscience 6407/10
Description
Fendiline Hydrochloride is a K-Ras inhibitor. Selectively inhibits K-Ras localization to the plasma membrane (IC50 = 9.64 μM). Blocks proliferation of cancer cell lines expressing oncogenic mutant K-Ras. Also inhibits L-type calcium channels (IC50 = 17 μM).Specifications
| Ras GTPase Inhibitors | |
| 13636-18-5 | |
| >98% | |
| 10 mg |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.