missing translation for 'onlineSavingsMsg'
Learn More

Fenoldopam Mesylate, TRC

Product Code. 30406959
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406959 5 mg
30406292 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30406959 Supplier Toronto Research Chemicals Supplier No. TRCF2488005MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67227-57-0
Molecular Formula C16 H16 Cl N O3 . C H4 O3 S
Molecular Weight (g/mol) 401.86
Synonym 1H-3-Benzazepine-7,8-diol, 6-chloro-2,3,4,5-tetrahydro-1-(4-hydroxyphenyl)-, methanesulfonate (1:1), 1H-3-Benzazepine-7,8-diol, 6-chloro-2,3,4,5-tetrahydro-1-(4-hydroxyphenyl)-, methanesulfonate (salt) (9CI), Corlopam, Fenoldopam mesylate, Fenoldopam methanesulfonate, Fenoldopam monomethanesulfonate, SKF 82526J
IUPAC Name 6-chloro-1-(4-hydroxyphenyl)-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol;methanesulfonic acid
SMILES CS(=O)(=O)O.Oc1ccc(cc1)C2CNCCc3c(Cl)c(O)c(O)cc23

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H16ClNO3.CH4O3S/c17-15-11-5-6-18-8-13(9-1-3-10(19)4-2-9)12(11)7-14(20)16(15)21;1-5(2,3)4/h1-4,7,13,18-21H,5-6,8H2;1H3,(H,2,3,4)
Formula Weight 401.07
Chemical Name or Material Fenoldopam Mesylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.