missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenpropathrin-d5, TRC
Specifications
Specifications
| Chemical Name or Material | Fenpropathrin-d5 |
| Molecular Formula | C22H18D5NO3 |
| Quantity | 10 mg |
| Synonym | 2,2,3,3-Tetramethyl-cyclopropanecarboxylic Acid Cyano(3-phenoxyphenyl)methyl Ester-d5, (±)-Fenpropathrin-d5, 2,2,3,3-Tetramethylcyclopropanecarboxylic acid cyano(3-phenoxyphenyl)methyl ester-d5, Danimen-d5, Danitol-d5, Danitol 10EC-d5, Danitol 2.4EC-d5, Danitol Fiori-d5, Fenpropanate-d5, Kilumal-d5, Meiothrin-d5, Meothrin-d5, Miothrin-d5, Rody-d5, S 3206-d5, SD 41706-d5, Smash-d5, Tame-d5, WL 41706-d5, XE 938-d5, α-Cyano-3-phenoxybenzyl 2,2,3,3-tetramethylcyclopropanecarboxylate-d5 |
| InChI Formula | InChI=1S/C22H23NO3/c1-21(2)19(22(21,3)4)20(24)26-18(14-23)15-9-8-12-17(13-15)25-16-10-6-5-7-11-16/h5-13,18-19H,1-4H3/i5D,6D,7D,10D,11D |
| SMILES | O=C(C1C(C)(C)C1(C)C)OC(C#N)C2=CC=CC(OC3=C([2H])C([2H])=C([2H])C([2H])=C3[2H])=C2 |
| Molecular Weight (g/mol) | 354.45 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?