missing translation for 'onlineSavingsMsg'
Learn More

Fenticonazole Nitrate, TRC

Product Code. 30403970
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403970 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403970 Supplier Toronto Research Chemicals Supplier No. TRCF2792501G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenticonazole nitrate
CAS 73151-29-8
Molecular Formula C24 H20 Cl2 N2 O S . H N O3
Quantity 1 g
Synonym Fenticonazole nitrate, 1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[4-(phenylsulfanyl)benzyl]oxy]ethyl]-1H-imidazole nitrate, 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-[[4-(phenylthio)phenyl]methoxy]ethyl]-, nitrate (1:1), 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-[[4-(phenylthio)phenyl]methoxy]ethyl]-, mononitrate (9CI), Falvin, Feniconazole nitrate, Fentiderm, REC 15-1476
InChI Formula InChI=1S/C24H20Cl2N2OS.HNO3/c25-19-8-11-22(23(26)14-19)24(15-28-13-12-27-17-28)29-16-18-6-9-21(10-7-18)30-20-4-2-1-3-5-20;2-1(3)4/h1-14,17,24H,15-16H2;(H,2,3,4)
SMILES O[N+](=O)[O-].Clc1ccc(C(Cn2ccnc2)OCc3ccc(Sc4ccccc4)cc3)c(Cl)c1
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(4-phenylsulfanylphenyl)methoxy]ethyl]imidazole;nitric acid
Molecular Weight (g/mol) 518.41
Formula Weight 517.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.