missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Buffer R (10X)

Product Code. 11531505 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 x 1.0 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11531505 5 x 1.0 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11531505 Supplier Thermo Scientific™ Supplier No. BR5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ensure the optimum reaction conditions for restriction enzymes with Thermo Scientific™ 10X Buffer R, premixed with BSA for enhanced stability.

Five Buffer System ensures the optimum reaction conditions for each restriction enzyme. The system consists of 10X B (blue), G (green), O (orange), R (red), and Tango (yellow) buffers. All restriction enzymes are supplied in color-coded tubes to indicate the recommended reaction buffer. The recommended buffer and/or the universal Tango buffer are supplied with each enzyme.

To ensure consistent enzyme performance, Thermo Scientific restriction enzyme buffers contain BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations. Multiple freeze-thaw cycles of the buffers will not cause BSA precipitation.

Restriction enzymes exhibit 100% of their certified activity in the recommended buffer. However, some enzymes require additives to achieve 100% activity. For example, AjuI, AlfI, BdaI, BplI, BseMII, FaqI, Eco57I, Eco57MI, Hin4I, and TsoI require S-adenosylmethionine, which is supplied with the enzyme, while AarI and BveI require an oligonucleotide (also supplied with the enzyme). Esp3I requires DTT.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Quantity 5 x 1.0 mL
Product Type Buffer R
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.