missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 5-Fluoroorotic Acid

Product Code. 10609920 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10609920 1 g 1g
10619920 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10609920 Supplier Thermo Scientific™ Supplier No. R0811
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Detect expression of the URA3 gene, which encodes orotine-5'-monophosphate (OMP) dicarboxylase with Thermo Scientific™ 5-Fluoroorotic Acid, used in yeast molecular genetics.

5-Fluoroorotic Acid is a yeast with an active URA3 gene (Ura+) converts 5-FOA to fluorodeoxyuridine, which is toxic to cells (see the scheme below). Yeast strains carrying a mutation in the URA3 gene grow in the presence of 5-FOA if the media is supplemented with uracil. 5-FOA is frequently used in experiments with Saccharomyces cerevisiae (URA3), Schizosaccharomyces pombe (URA4 and URA5), Candida albicans (URA3), and E. coli (pyrF).

Yeast with an active URA3 gene (Ura+) converts 5-FOA to fluorodeoxyuridine, which is toxic to cells (see the scheme below). Yeast strains carrying a mutation in the URA3 gene grow in the presence of 5-FOA if the media is supplemented with uracil. 5-FOA is frequently used in experiments with Saccharomyces cerevisiae (URA3), Schizosaccharomyces pombe (URA4 and URA5), Candida albicans (URA3) and E.coli (pyrF).

Recommended for:

Detection of URA3 gene expression in yeast; Construction of yeast 2-hybrid libraries (1); Other areas of yeast genetics including gene replacement, analysis of transposable genetic elements and identification of gene mutations.

Note:

Low solubility in water. Solubility in dimethylsulfoxide is greater than 150mg/mL. Preparation of a 100mg/mL solution in dime-thylsulfoxide is recommended.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Fluoroorotic Acid
Product Type 5-Fluoroorotic Acid
Quantity 1 g
Form Powder
compliance-icons
Product Identifier
  • 5-Fluoroorotic acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 5-Fluoroorotic acidmonohydrate

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.