missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNA Loading Dye & SDS Solution (6X)

Product Code. 11571575 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11571575 5 x 1 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11571575 Supplier Thermo Scientific™ Supplier No. R1151
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform agarose gel analysis of DNA samples that contain high amounts of DNA binding proteins with this DNA loading dye containing SDS.

6X DNA Loading Dye & SDS Solution is recommended for agarose gel analysis of DNA samples that contain high amounts of DNA binding proteins. The presence of SDS eliminates DNA-protein interactions and prevents gel-shifts. The high concentration of EDTA protects DNA from degradation by metal-dependent nucleases.

Highlights

  • 1% SDS eliminates DNA-protein interactions, prevents appearance of additional bands due to annealing of DNA molecules with cohesive ends
  • 100 mM EDTA inhibits metal-dependent nucleases
TRUSTED_SUSTAINABILITY

Specifications

Concentration 6X
Gel Compatibility Acrylamide Gels, Agarose Gels
Label or Dye Bromophenol Blue, Xylene Cyanol FF (XCFF)
Product Type DNA Loading Dye & SDS Solution
Quantity 5 x 1 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.