missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lambda DNA (dam–, dcm–)

Product Code. 10252690 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10252690 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10252690 Supplier Thermo Scientific™ Supplier No. SD0021
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Generate DNA size marker fragments with this linear, double-stranded DNA, a common substrate for restriction endonucleases.

Lambda is a temperate Escherichia coli bacteriophage. The virion DNA is linear and double-stranded (48502 nt) with 12 bp single-stranded complementary 5'-ends. After the phage particle injects its chromosome into the cell, the chromosome circularizes by end joining. In the lytic pathway, phage genes encoding replication, lysis, and virion proteins are expressed. The chromosome replicates, and the replicas are cleaved and packaged into progeny phage particles. In the lysogenic pathway, phage gene expression is repressed, and the circular chromosome inserts into the bacterial chromosome by recombination.

Highlights

Phage lambda DNA is a common substrate for restriction endonucleases and for generating DNA size marker fragments. For large scale isolation of phage DNA, cI857Sam7, a mutant carrying four known mutations, is often used. The DNA sequence used to construct a phage alpha restriction map includes these mutations. The map shows enzymes that cut lambda DNA. Enzymes produced by Thermo Scientific are shown in orange (see Figure bellow).

Lambda DNA (dam-, dcm-) is isolated from a heat-inducible lysogenic E. coli GM2163 strain.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.3 μg/μL
Sample Type DNA
Final Product Type DNA
For Use With (Application) CloningNucleic Acid Gel Electrophoresis
Includes [other]
Product Type Lambda DNA
Quantity 500 μg
Species Lambda phage

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.