missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ dITP Solution (100 mM)

Product Code. 10681041 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
0.25 mL
Unit Size:
0.25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10681041 0.25 mL 0.25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10681041 Supplier Thermo Scientific™ Supplier No. R1191
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific dITP (2'-deoxyinosine 5'-triphosphate) is a highly stable nucleotide supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH.

Thermo Scientific dITP (2'-deoxyinosine 5'-triphosphate) is a highly stable nucleotide supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. The nucleotide has greater than 99% purity and is free of nuclease activities and human, and E. coli DNA. Thermo Scientific dITP is designed for use with a wide variety of different molecular biology applications.

Applications
• PCR and RT-PCR with Taq DNA polymerase (Cat. No. EP0072)
• qPCR
• cDNA synthesis
• SNP genotyping
• Primer extension.

Note: dITP is not suitable for PCR with proofreading enzymes or enzyme mixes containing proofreading enzymes.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100 mM
Content And Storage dITP Solution, 100 mM (0.25 mL)

Store at –20°C.
Form Liquid
Polymerase DNA Polymerase
Label or Dye Unlabeled
Quantity 0.25 mL
Shipping Condition Dry Ice
For Use With (Application) Standard PCR, cDNA Synthesis, RT-PCR

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.